missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IGFBP-2 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 3 publications
Brand: R&D Systems AF674
This item is not returnable.
View return policy
Description
IGFBP-2 Polyclonal specifically detects IGFBP-2 in Human samples. It is validated for Western Blot, Simple Western, Neutralization.
Specifications
| IGFBP-2 | |
| Polyclonal | |
| Unconjugated | |
| CAA34373 | |
| IGFBP2 | |
| Mouse myeloma cell line NS0-derived recombinant human IGFBP-2 Glu40-Gln328 Accession # CAA34373 | |
| 100 μg | |
| Primary | |
| Detects human IGFBP-2 in direct ELISAs and Western blots. | |
| Human | |
| IgG |
| Western Blot, Neutralization | |
| LYOPH | |
| Western Blot 0.5 ug/mL, Simple Western 25 ug/mL, Neutralization 2.5-7.5 ug/mL | |
| BP2, IBP2, IBP-2, IGF-binding protein 2, IGFBP2, IGFBP-2, IGF-BP53, insulin-like growth factor binding protein 2 (36kD), insulin-like growth factor binding protein 2, 36kDa, insulin-like growth factor-binding protein 2 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3485 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction