missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human IGF-II Recombinant Protein, PeproTech®
Click to view available options
Quantity:
1 mg
10 μg
100 μg
250 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
250µg
500µg
50µg
Description
Recombinant Human IGF-I and IGF-II are globular proteins containing 70 and 67 amino acids, respectively, and 3 intra-molecular disulfide bonds. The calculated molecular weight of Recombinant Human IGF-II is 7.5 kDa. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
The IGFs are mitogenic, polypeptide growth factors that stimulate the proliferation and survival of various cell types, including muscle, bone, and cartilage tissue in vitro. IGFs are predominantly produced by the liver, although a variety of tissues produce the IGFs at distinctive times. The IGFs belong to the Insulin gene family, which also contains insulin and relaxin. The IGFs are similar to insulin by structure and function, but have a much higher growth-promoting activity than insulin. IGF-II expression is influenced by placenta lactogen, while IGF-I expression is regulated by growth hormone. Both IGF-I and IGF-II signal through the tyrosine kinase type I receptor (IGF-IR), but IGF-II can also signal through the IGF-II/Mannose-6-phosphate receptor. Mature IGFs are generated by proteolytic processing of inactive precursor proteins, which contain N-terminal and C-terminal propeptide regions.
Specifications
Specifications
| Accession Number | P01344 |
| For Use With (Application) | ELISA, Functional Assay, Western Blot |
| Formulation | protein with no preservative |
| Gene ID (Entrez) | 3481 |
| Molecular Weight (g/mol) | 7.5 kDa |
| Name | Human IGF-II |
| Quantity | 100 μg |
| Source | E. coli |
| Regulatory Status | RUO |
| Endotoxin Concentration | <1 EU/ μg |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction