missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IGF-II R/IGF2R Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems BAF2447
This item is not returnable.
View return policy
Description
IGF-II R/IGF2R Polyclonal specifically detects IGF-II R/IGF2R in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA Detection (Matched Antibody Pair).
Specifications
| IGF-II R/Mannose 6 Phosphate Receptor (Cation independent) | |
| Polyclonal | |
| Biotin | |
| P11717 | |
| IGF2R | |
| Mouse myeloma cell line NS0-derived recombinant human IGF-II R Ser1510-Phe2108 Accession # P11717 | |
| 50 μg | |
| Primary | |
| Detects human IGF-II R in ELISAs and Western blots. In sandwich ELISAs, less than 0.2% cross-reactivity with recombinant human (rh) IGF-I R, rhIGF-I, and rhIGF-II is observed. | |
| Human | |
| IgG |
| Western Blot, Flow Cytometry, ELISA | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL | |
| 300 kDa mannose 6-phosphate receptor, cation-independent mannose-6 phosphate receptor, cation-independent mannose-6-phosphate receptor, CD222, CD222 antigen, CI Man-6-P receptor, CIMPR, CI-MPR, IGF2R, IGF-II receptor, IGFIIR, IGF-IIR, insulin-like growth factor 2 receptorM6P/IGF2R, Insulin-like growth factor II receptor, M6P/IGF2 receptor, M6PR, M6P-R, MPR 300, MPR1, MPRI, MPRIM6PR | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3482 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction