missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IGF-II R/IGF2R Antibody, R&D Systems™

Mouse Monoclonal Antibody
£167.00 - £501.00
Specifications
| Antigen | IGF-II R/Mannose 6 Phosphate Receptor (Cation independent) |
|---|---|
| Clone | 372616 |
| Dilution | ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18701678
![]() |
R&D Systems
MAB24471-SP |
25 μg |
£167.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18191471
|
R&D Systems
MAB24471 |
500 μg |
£501.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IGF-II R/IGF2R Monoclonal specifically detects IGF-II R/IGF2R in Human samples. It is validated for ELISA Capture (Matched Antibody Pair).Specifications
| IGF-II R/Mannose 6 Phosphate Receptor (Cation independent) | |
| ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 300 kDa mannose 6-phosphate receptor, cation-independent mannose-6 phosphate receptor, cation-independent mannose-6-phosphate receptor, CD222, CD222 antigen, CI Man-6-P receptor, CIMPR, CI-MPR, IGF2R, IGF-II receptor, IGFIIR, IGF-IIR, insulin-like growth factor 2 receptorM6P/IGF2R, Insulin-like growth factor II receptor, M6P/IGF2 receptor, M6PR, M6P-R, MPR 300, MPR1, MPRI, MPRIM6PR | |
| IGF2R | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 372616 | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Human | |
| P11717 | |
| 3482 | |
| Mouse myeloma cell line NS0-derived recombinant human IGF-II R Ser1510-Phe2108 Accession # P11717 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human IGF-II R in ELISAs. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title