missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human IGF-II/IGF2 DuoSet ELISA
Click to view available options
Quantity:
1 Kit (5 Plates)
Unit Size:
Pack of 5
Description
The Human IGF-II/IGF2 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human IGF-II/IGF2.
Specifications
Specifications
| Assay Range | 23.40 - 1500pg/mL |
| Target Species | Human |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 3481 |
| Quantity | 1 Kit (5 Plates) |
| Storage Requirements | Store the unopened product at 2° to 8°C. Do not use past expiration date. |
| Synonym | C11orf43, chromosome 11 open reading frame 43, FLJ22066, FLJ44734, GRDF, IGF2, IGF-2, IGFII, IGF-II, insulin-like growth factor 2 (somatomedin A), insulin-like growth factor II, insulin-like growth factor type 2, MSA, PEG2, PP9974, somatomedin-A |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction