missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IGF-I/IGF-1 Antibody, R&D Systems™

Goat Monoclonal Antibody
Brand: R&D Systems MAB2911
This item is not returnable.
View return policy
Description
IGF-I/IGF-1 Monoclonal specifically detects IGF-I/IGF-1 in Human samples. It is validated for Immunohistochemistry.
Specifications
| IGF-I/IGF-1 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| IBP1, IGF1, IGF-1, IGF1A, IGFI, IGF-I, IGF-IA, IGF-IB, insulin-like growth factor 1, insulin-like growth factor 1 (somatomedin C), insulin-like growth factor I, insulin-like growth factor IA, insulin-like growth factor IB, Mechano growth factor, MGF, Somatomedin A, Somatomedin C, Somatomedin-C | |
| Goat | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 3479 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG |
| Immunohistochemistry | |
| 40017A | |
| Immunohistochemistry 3-25 ug/mL | |
| P05019 | |
| IGF1 | |
| E. coli-derived recombinant human IGF-I/IGF-1, Gly49-Ala118, Accession # P05019 | |
| 100 μg | |
| Primary | |
| Detects human IGF-I/IGF-1 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse IGF-I, recombinant rat IGF-I or recombinant human IGF-II is observed. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction