missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human IGF-I, Animal-Free Recombinant Protein, PeproTech®

Product Code. 17823253
Change view
Click to view available options
Quantity:
1 mg
100 μg
500 μg
Unit Size:
100µg
1mg
500µg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17823253 1 mg 1mg
17863643 100 μg 100µg
17853303 500 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17823253 Supplier Gibco™ Supplier No. AF100111MG

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Recombinant Protein

Recombinant Human IGF-I and IGF-II are globular proteins containing 70 and 67 amino acids, respectively, and 3 intra-molecular disulfide bonds. The calculated molecular weight of Recombinant Human IGF-I is 7.6 kDa. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

IGF1 (Insulin-like growth factor-1) is structurally and functionally related to insulin but has a much higher growth-promoting activity. A variety of cellular responses are induced by IGF1, including cell proliferation, differentiation, migration, and survival. Further, IGF1 is a polypeptide growth factor that stimulates the proliferation of a wide range of cell types in muscle, bone, and cartilage tissue. IGF1 stimulates glucose transport in rat bone-derived osteoblastic (PyMS) cells and is effective at much lower concentrations than insulin, not only regarding glycogen and DNA synthesis but also with regards to enhancing glucose uptake. In circulation, IGFs are predominantly bound to binding proteins (IGFBPs) which prolong the half-life of the IGFs and play a role in delivering them to target tissues. IGF-I is known as one of the most potent activators of the AKT signaling pathway which is known to be a stimulator of proliferation and an inhibitor of programmed cell death. Moreover, mature human IGF-I is 100% homologous with bovine and porcine proteins. Low levels of IGF1 have been linked to Alzheimer's disease. IGF1 is processed from a precursor, bound by a specific receptor, and secreted. Defects in the IGF1 gene are a cause of insulin-like growth factor 1 deficiency and several transcript variants encoding different isoforms have been found.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P05019
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 3479
Molecular Weight (g/mol) 7.6 kDa
Name Human IGF-I, Animal-Free
Quantity 1 mg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <0.1 EU/ μg
Gene Alias C730016P09Rik; class 1 insulin-like growth factor I preproprotein; class 2 insulin-like growth factor I preproprotein; H-IGF-1; IBP1; IGF; IGF1; Igf-1; IGF1A; IGFI; Igf-I; IGFIa; IGF-IA; IGF-IB; insulin growth factor-1; Insulin like growth factor; insulin like growth factor 1; insulin-like growth factor 1; insulin-like growth factor 1 (somatomedin C); insulin-like growth factor I; insulin-like growth factor I (somatomedin C); insulin-like growth factor IB; mechano growth factor; MGF; M-IGF-1; OTTHUMP00000195084; prepro-IGF-I; prepro-insulin-like growth factor I; R-IGF-1; somatomedin; Somatomedin C; Somatomedin-C
Common Name IGF1
Gene Symbol IGF1
Biological Activity The ED50 was determined by a cell proliferation assay using FDC-P1 cells is ≤ 2.0 ng/ml, corresponding to a specific activity of ≥ 5 x 10^5 units/mg
Conjugate Unconjugated
Recombinant Recombinant
Sequence GPETLCGAEL VDALQFVCGD RGFYFNKPTG YGSSSRRAPQ TGIVDECCFR SCDLRRLEMY CAPLKPAKSA
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.