missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Human IgA ELISA Kit (Chemiluminescence)

Product Code. 18382205 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18382205 1 Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18382205 Supplier Novus Biologicals™ Supplier No. NBP300397

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human IgA ELISA Kit (Chemiluminescence) measures IgA in Serum, plasma, cell supernatant, and other biological fluids.

TRUSTED_SUSTAINABILITY

Specifications

Assay Range 3.13 to 200 ng/mL
Assay Sensitivity 1.88 ng/mL
Product Type ELISA Kit (Chemiluminescence)
Sample Type Cell Culture Supernatant, Other Biological Fluids, Plasma, Serum
Specificity This kit recognizes Human IgA in samples. No significant cross-reactivity or interference between Human IgA and analogues was observed.
Target Species Human
For Use With (Application) ELISA
Gene ID (Entrez) 3493
Gene Symbol IGHA1
Quantity 1 Kit
Research Discipline Adaptive Immunity, Immunology
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym Immunoglobulin A
Target IgA
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.