missing translation for 'onlineSavingsMsg'
Learn More

Human IgA Antibody, R&D Systems™
GREENER_CHOICE

Product Code. 18770779
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18770779 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18770779 Supplier R&D Systems Supplier No. AF4787100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Antibody

IgA specifically detects IgA in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IgA
Applications Western Blot
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene Alias Immunoglobulin A
Host Species Sheep
Immunogen Purified human IgA from pooled normal Human colostrum
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 3493
Test Specificity Detects human IgA in Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.