missing translation for 'onlineSavingsMsg'
Learn More

Human IgA Antibody (BFG-267), Alexa Fluor™ 594, Novus Biologicals™

Product Code. 18405643 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.2 mL
Unit Size:
0.2mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18405643 0.2 mL 0.2mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18405643 Supplier Novus Biologicals Supplier No. NBP142270AF594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Human IgA Monoclonal antibody specifically detects Human IgA in Human samples. It is validated for ELISA, Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen Human IgA
Applications ELISA, Immunohistochemistry
Classification Monoclonal
Clone BFG-267
Conjugate Alexa Fluor 594
Dilution ELISA, Immunohistochemistry
Formulation 50mM Sodium Borate
Gene Alias CD79A;IGA;Igalpha;Ig-alpha;Ly54;Ly-54;MB1;MB-1
Host Species Mouse
Immunogen Purified alpha-chain (Fc) of human IgA
Purification Method Ion exchange chromatography
Quantity 0.2 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.