missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Ig Lambda Light Chain Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB10049-SP
This item is not returnable.
View return policy
Description
Human lambda light chain Monoclonal specifically detects Human lambda light chain in Human samples. It is validated for Western Blot, Simple Western, ELISA.
Specifications
| Human lambda light chain | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG1 |
| Western Blot, ELISA | |
| 1000011 | |
| Western Blot 2 μg/mL, Simple Western 20 μg/mL, ELISA | |
| Bence Jones protein, BJP, Constant region of lambda light chains, HCAK 1, HCAK1, Ig kappa chain C region, Ig lambda chain C regions, IGKC, IGLC, IGLC 1, IGLC 2, IGLC 3, IGLC1, IGLC2, IGLC3, Immunoglobulin kappa constant, Immunoglobulin kappa constant region, Immunoglobulin kappa light chain, Immunoglobulin lambda constant 1, Kappa 1 immunoglobulin light chain, Km, Mcg marker, MGC111575, MGC62011, MGC72072, MGC88770, MGC88771, MGC88809, Paraprotein | |
| Ig Lambda Light Chain | |
| 25 μg | |
| Detects Human Ig Lambda Light Chain in direct ELISAs. | |
| Human | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction