missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Ig kappa light chain Rabbit anti-Human, Clone: RM126 (Biotin), Abnova™
Rabbit Monoclonal Antibody
Brand: Abnova MAB14949.50ug
This item is not returnable.
View return policy
Description
Rabbit monoclonal antibody raised against native human Ig kappa light chain.
- High specificity
- High reactivity
Specifications
| Human Ig kappa light chain | |
| Monoclonal | |
| Biotin | |
| ELISA (0.05-0.2 ug/mL) Flow Cytometry Immunocytochemistry (0.5-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-2 ug/mL) The optimal working dilution should be determined by the end user. | |
| P01834 | |
| IGKC | |
| Native purified human IgG. | |
| 50 μg | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (PFA fixed) | |
| RM126 | |
| Rabbit monoclonal antibody raised against native human Ig kappa light chain. | |
| In PBS (50% glycerol, 1% BSA, 0.09% sodium azide) | |
| HCAK1/Km/MGC111575/MGC62011/MGC72072/MGC88770/MGC88771/MGC88809 | |
| Rabbit | |
| Protein A purification | |
| RUO | |
| 3514 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction