missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human IFT122 (aa 751-835) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (91%), Rat (91%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-59742 (PA5-59742. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
This gene encodes a member of the WD repeat protein family. WD repeats are minimally conserved regions of approximately 40 amino acids typically bracketed by gly-his and trp-asp (GH-WD), which may facilitate formation of heterotrimeric or multiprotein complexes. Members of this family are involved in a variety of cellular processes, including cell cycle progression, signal transduction, apoptosis, and gene regulation. This cytoplasmic protein contains seven WD repeats and an AF-2 domain which function by recruiting coregulatory molecules and in transcriptional activation. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
Specifications
Specifications
| Accession Number | Q9HBG6 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 55764 |
| Name | Human IFT122 (aa 751-835) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | C86139; CED; CED1; hypothetical protein zgc:77217; Ift122; intraflagellar transport 122; intraflagellar transport 122 homolog; intraflagellar transport 122 homolog (Chlamydomonas); intraflagellar transport protein 122 homolog; sopb; SPG; WD repeat domain 10; WD repeat-containing protein 10; WD repeat-containing protein 140; WDR10; WDR10p; WDR140; zgc:77217 |
| Common Name | IFT122 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction