missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IFN-kappa Antibody, R&D Systems™

Mouse Monoclonal Antibody
£156.00 - £387.00
Specifications
| Antigen | IFN-kappa |
|---|---|
| Clone | 1009727 |
| Dilution | ELISA |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18721597
![]() |
R&D Systems
MAB102472-SP |
25 μg |
£156.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18781477
![]() |
R&D Systems
MAB102472-100 |
100 μg |
£387.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IFN-kappa Monoclonal specifically detects IFN-kappa in Human samples. It is validated for ELISA.Specifications
| IFN-kappa | |
| ELISA | |
| Monoclonal | |
| Mouse | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| IFNK, IFNkappa, IFN-kappa | |
| E. coli-derived recombinant human IFN-kappa , Leu28-Lys207, Accession # Q90W0 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human IFN-kappa in direct ELISAs. |
| 1009727 | |
| ELISA | |
| Unconjugated | |
| Human | |
| Q90W0 | |
| 56832 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title