missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human IFN-gamma XL Luminex™ Performance Assay
Click to view available options
Quantity:
1 Kit
Unit Size:
Each
Specifications
Specifications
| Quantity | 1 Kit |
| Species | Human |
| Gene ID (Entrez) | 3458 |
| Synonym | IFG, IFI, IFNG, IFNgamma, IFN-gamma, Immune interferon, interferon gamma, interferon, gamma |
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction