Learn More
Invitrogen™ IFN gamma Human ELISA Kit, High Sensitivity
Description
Includes
- Aluminum pouch(es) with a Microwell Plate coated with monoclonal antibody to human IFN-gamma
- Biotin-Conjugate anti-human IFN-gamma monoclonal antibody
- Streptavidin-HRP
- Human IFN-gamma Standard lyophilized, 200ng/mL upon reconstitution
- Sample Diluent
- Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
- Amplification Diluent Concentrate (2x)
- Amplification Reagent I
- Amplification Reagent II
- Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
- Substrate Solution
- Stop Solution (1M Phosphoric acid)
- Adhesive Films
The Human High Sensitivity Interferon gamma (Hu HS IFNγ) ELISA quantitates Hu HS IFNγ in human serum, plasma, buffered solution, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu HS IFNγ. Principle of the method The Human HS IFNγ solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. A special amplification reagent step is added to achieve the high sensitivity capability. After incubation and washing steps to rid the microplate of unbound substances, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Specifications
Specifications
| Accession Number | P01579 |
| Assay Range | 0.16-10.0 pg/mL |
| Assay Sensitivity | 0.06 pg/mL |
| Conjugate | Biotin |
| Product Type | ELISA Kit |
| Sample Type | Plasma, Serum, Supernatant |
| Target Species | Human |
| For Use With (Application) | ELISA |
| For Use With (Equipment) | Colorimetric Microplate Reader |
| Gene ID (Entrez) | 3458 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.