missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IFN-gamma F(ab')2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£176.00 - £423.00
Specifications
| Antigen | IFN-gamma |
|---|---|
| Clone | 25723R-FAB2 |
| Dilution | Intracellular Staining by Flow Cytometry 0.25 ug/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18740417
![]() |
R&D Systems
MAB285R-FAB2-SP |
25 μg |
£176.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18782177
![]() |
R&D Systems
MAB285R-FAB2-100 |
100 μg |
£423.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IFN-gamma Monoclonal antibody specifically detects IFN-gamma in Human samples. It is validated for Flow CytometrySpecifications
| IFN-gamma | |
| Intracellular Staining by Flow Cytometry 0.25 ug/mL | |
| Monoclonal | |
| Mouse | |
| Human | |
| AAP20098.1 | |
| 3458 | |
| IgG2b | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
| 25723R-FAB2 | |
| Flow Cytometry | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| IFG, IFI, IFNG, IFNgamma, IFN-gamma, Immune interferon, interferon gamma, interferon, gamma | |
| E. coli-derived recombinant human IFN-gamma , Gln24-Gln166, Accession # AAP20098.1 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title