Learn More
Invitrogen™ IFN gamma Human Uncoated ELISA Kit with Plates
Specifically engineered for accurate and precise measurement of human IFN gamma protein levels from samples including serum, plasma, and cell culture supernatants.
£165.00 - £546.00
Specifications
Accession Number | P01579 |
---|---|
Assay Range | 4-500 pg/mL |
Assay Sensitivity | 4 pg/mL |
Conjugate | Biotin |
Product Type | ELISA Kit |
Alert:
Contains Azide
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15521107
|
Invitrogen™
88-7316-22 |
2 x 96 Tests |
£165.00
Pack of 2 |
Estimated Shipment: 05-04-2024
Log in to see stock. |
|||||
15551107
|
Invitrogen™
88-7316-86 |
10 x 96 Tests |
£392.00
Pack of 10 |
In Stock
Log in to see stock. |
|||||
15531107
|
Invitrogen™
88-7316-76 |
20 x 96 Tests |
£546.00
Pack of 20 |
In Stock
Log in to see stock. |
|||||
Description
The Human Interferon gamma (IFNγ) Uncoated ELISA Kit contains pre-matched antibody pairs, plates and reagents for performing quantitative enzyme linked immunosorbent assays (ELISA) to detect and quantify protein levels of human IFNγ. Wash Buffer and Stop Solution are needed to complete the ELISA reaction and are sold separately. Principle of the method ELISAs are designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody is coated to the bottom of the wells of a microplate, which is an overnight process. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. A sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.
Human IFN gamma ELISA Ready-SET-Go! reagent set (with or without high-affinity binding microwell plates) contains the necessary reagents, buffers and diluents for performing quantitative enzyme linked immunosorbent assays (ELISA)- To ensure optimal results from the Human IFN gamma ELISA Ready-SET-Go! Set, please only use the components included in the set. Exchanging of components may lead to a change in signal
Specifications
P01579 | |
4 pg/mL | |
ELISA Kit | |
Human | |
Colorimetric Microplate Reader | |
CSIF,IL-10 | |
HRP | |
Plasma, 100 μL; Serum, 100 μL; Supernatant, 100 μL | |
Interferon-gamma, IFN-g, IFNg | |
1 hr. 45 min. |
4-500 pg/mL | |
Biotin | |
Plasma, Serum, Supernatant | |
ELISA | |
3458 | |
Capture Antibody: Pre-titrated; purified antibody, Detection Antibody: Pre-titrated; biotin-conjugated antibody, Standard: Recombinant protein for generating standard curve and calibrating samples, 10X Coating Buffer: Buffer for plating the Capture Antibody, 5X ELISA/ELISPOT Diluent: Buffer for blocking and diluting the Detection Antibody and Enzyme, Enzyme: Pre-titrated Avidin-HRP, Substrate: 1X TMB Solution, Certificate of Analysis: Lot-specific instructions for dilution of antibodies and standards, 96 Well Plate: Corning Costar 9018 (included with product Cat. Nos. ending in suffixes -22; -76; -86) | |
RUO | |
2°C to 8°C | |
Human | |
24 hr. 30 min. |
For Research Use Only