Learn More
STEMCELL Technologies Human IFN-gamma ELISA Kit
Description
- The Human Interferon-gamma (IFN-γ) ELISA Kit is designed for the quantitative detection and measurement of human IFN-γ in biological fluids such as serum, plasma, and cell culture supernatants
- IFN-γ is primarily secreted by T and NK cells
- IFN-γ is a pleiotropic cytokine that modulates a broad range of immunological functions including antimicrobial and antitumor activity, and increases the surface expression of MHC class I and II proteins
- The assay is based on the sandwich ELISA method, in which samples are added to ELISA strip plates pre-coated with capture antibodies specific for the cytokine
- The captured cytokine is detected by addition of a biotinylated detection antibody, followed by streptavidin-horseradish peroxidase, which binds the biotinylated antibody
- Addition of the chromogenic enzyme substrate 3,3',5,5' tetramethylbenzidine (TMB) results in a colored product with an intensity directly proportional to the concentration of cytokine in the sample
Specifications
Specifications
| Description | Dendritic Cells; Macrophages; NK Cells; T Cells |
| Quantity | 2 Plates |