Learn More
Human IFN-gamma Antibody, R&D Systems™
GREENER_CHOICE
Rabbit Monoclonal Antibody
Brand:  R&D Systems MAB2853-SP
 View more versions of this product
Product Code. 18774577
  • £170.00 / 25µg
Description
IFN-gamma Monoclonal specifically detects IFN-gamma in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IFN-gamma
Monoclonal
Unconjugated
Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS.
IFG, IFI, IFNG, IFNgamma, IFN-gamma, Immune interferon, interferon gamma, interferon, gamma
E. coli-derived recombinant human IFN-gamma , Gln24-Gln166, Accession # AAP20098.1
25 μg
Primary
Detects human IFN-gamma in direct ELISAs.
Human
Purified
Immunohistochemistry, Immunocytochemistry
2485E
Immunohistochemistry 5-25 ug/mL, Immunocytochemistry 5-25 ug/mL
AAP20098.1
Rabbit
Protein A or G purified from cell culture supernatant
RUO
3458
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.