missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IFN-gamma Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 15 publications
Brand: R&D Systems MAB2852-500
This item is not returnable.
View return policy
Description
IFN-gamma Monoclonal specifically detects IFN-gamma in Human samples. It is validated for Western Blot, Neutralization, Intracellular Staining by Flow Cytometry, ELISA Capture (Matched Antibody Pair).
Specifications
| IFN-gamma | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS and NaCl with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| IFNG | |
| E. coli-derived recombinant human IFN-gamma | |
| 500 μg | |
| Primary | |
| Detects human IFN-gamma in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant mouse IFN-gamma , recombinant rat IFN-gamma , recombinant porcine IFN-gamma , recombinant feline IFN-gamma , or recombinant cotton rat IFN-gamma is observed. | |
| Human | |
| Supernatant |
| Western Blot, Neutralization, ELISA | |
| K3.53 | |
| Western Blot 1 ug/mL, Neutralization ≤2 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| IFG, IFI, IFNG, IFNgamma, IFN-gamma, Immune interferon, interferon gamma, interferon, gamma | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3458 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction