missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IFN-beta Antibody, R&D Systems™

Description
IFN-beta Monoclonal specifically detects IFN-beta in Human samples. It is validated for ELISA.
Specifications
Specifications
| Antigen | IFN-beta |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 944411 |
| Conjugate | Unconjugated |
| Dilution | ELISA |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. |
| Gene Accession No. | P01574 |
| Gene Alias | Fibroblast interferon, IFB, IFBIFNB, IFF, IFNB, IFNB1, IFNbeta, IFN-beta, interferon beta, interferon, beta 1, fibroblast, MGC96956 |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?