missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IFN-beta Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems MAB8142-SP
This item is not returnable.
View return policy
Description
IFN-beta Monoclonal specifically detects IFN-beta in Human samples. It is validated for Immunocytochemistry.
Specifications
| IFN-beta | |
| Monoclonal | |
| Unconjugated | |
| Supplied as a solution in PBS containing BSA, Glycerol and Sodium Azide. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide | |
| Fibroblast interferon, IFB, IFBIFNB, IFF, IFNB, IFNB1, IFNbeta, IFN-beta, interferon beta, interferon, beta 1, fibroblast, MGC96956 | |
| Chinese hamster ovary cell line CHO-derived recombinant human IFN-beta Met1-Asn187 Accession # P01574 | |
| 25 μg | |
| Primary | |
| Detects human IFN-beta in direct ELISAs. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG |
| Flow Cytometry, Immunocytochemistry | |
| 2036A | |
| Flow Cytometry 0.25 uL/10^6 cells, Immunocytochemistry 1-25 ug/mL | |
| P01574 | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 3456 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction