Learn More
Human IFN-beta Antibody, R&D Systems™
GREENER_CHOICE
Rabbit Monoclonal Antibody
Brand:  R&D Systems MAB8142-100
 View more versions of this product
Product Code. 18750148
  • £428.00 / 100µg
Description
IFN-beta Monoclonal specifically detects IFN-beta in Human samples. It is validated for Immunocytochemistry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IFN-beta
Monoclonal
Unconjugated
Supplied as a solution in PBS containing BSA, Glycerol and Sodium Azide. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide
Fibroblast interferon, IFB, IFBIFNB, IFF, IFNB, IFNB1, IFNbeta, IFN-beta, interferon beta, interferon, beta 1, fibroblast, MGC96956
Chinese hamster ovary cell line CHO-derived recombinant human IFN-beta Met1-Asn187 Accession # P01574
100 μg
Primary
Detects human IFN-beta in direct ELISAs.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
IgG
Flow Cytometry, Immunocytochemistry
2036A
Flow Cytometry 0.25 uL/10^6 cells, Immunocytochemistry 1-25 ug/mL
P01574
Rabbit
Protein A or G purified from cell culture supernatant
RUO
3456
Human
Purified