missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human IFN-alpha A/D Recombinant Protein

Human IFN-alpha A/D Recombinant Protein

Brand:  Gibco™ PHC4044

Code : 50

Additional Details : Weight : 0.02000kg

Product Code. 10334263

  • £417.00 / Each
Estimated Shipment: 17-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


For Research Use Only. All usage must comply with product instructions.
 

Description

Description

Human IFN-alpha A/D Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN

IFN-α is involved in anti-viral and anti-parasitic activitites.

Shipping Condition: Dry Ice.

Specifications

Specifications

Cell Culture
19.4 kDa
IFN-α A/D
Store in ultra-cold freezer (-68 to -85°C).
Dry Ice
Cytokines & Receptors
Liquid
Immunology, Virology, Oncology, Signal Transduction
95 %
3438
1 x 10 exp. 5 units
Human
10^5 units
E. coli
Recombinant, Ligand
Interferons
Gibco™
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.