missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human IFITM1 Synthetic Peptide
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
Description
IFITM1 Synthetic Peptide, PEP-1102, from Invitrogen.
IFITM1 (Interferon inducible transmembrane protein 1) is a member of the IFN-inducible transmembrane protein family. It is an essential mediator of interferon-gamma-induced antiproliferation and plays a role in the control of cell growth. IFITM1 is upregulated in several tumor types and may be useful as a tumor biomarker. Both mouse IFITM1 and IFITM3 are expressed on the cell surfaces of primordial germ cells in a developmentally-regulated manner. IFITM1 activity is required for primordial germ cell transit, and IFITM1 acts as a repulsive molecule by repelling non-IFITM1-expressing primordial germ cells from the mesoderm into the endoderm.
Specifications
Specifications
| Gene ID (Entrez) | 8519 |
| Species | Human |
| Purification Method | purified |
| Accession Number | P13164 |
| Common Name | Human IFITM1 |
| Concentration | 200 μg/mL |
| Content And Storage | -20°C |
| Description | Synthetic Peptide |
| Format | Liquid |
| Formulation | PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction