missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human IDH3G (aa 267-380) Control Fragment Recombinant Protein

Product Code. 30195797
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30195797

Brand: Invitrogen™ RP91985

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (98%), Rat (98%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-51385 (PA5-51385. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Isocitrate dehydrogenases catalyze the oxidative decarboxylation of isocitrate to 2-oxoglutarate. These enzymes belong to two distinct subclasses, one of which utilizes NAD(+) as the electron acceptor and the other NADP(+). Five isocitrate dehydrogenases have been reported: three NAD(+)-dependent isocitrate dehydrogenases, which localize to the mitochondrial matrix, and two NADP(+)-dependent isocitrate dehydrogenases, one of which is mitochondrial and the other predominantly cytosolic. NAD(+)-dependent isocitrate dehydrogenases catalyze the allosterically regulated rate-limiting step of the tricarboxylic acid cycle. Each isozyme is a heterotetramer that is composed of two alpha subunits, one beta subunit, and one gamma subunit. The protein encoded by this gene is the gamma subunit of one isozyme of NAD(+)-dependent isocitrate dehydrogenase. This gene is a candidate gene for periventricular heterotopia. Several alternatively spliced transcript variants of this gene have been described, but only some of their full length natures have been determined.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P51553
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 3421
Name Human IDH3G (aa 267-380) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias H-IDHG; IDH; IDH3G; IDH-gamma; iso; Isocitrate dehydrogenase [NAD] subunit gamma 1, mitochondrial; isocitrate dehydrogenase [NAD] subunit gamma, mitochondrial; isocitrate dehydrogenase 3 (NAD(+)) gamma; isocitrate dehydrogenase 3 (NAD), gamma; isocitrate dehydrogenase 3 (NAD+) gamma; isocitrate dehydrogenase 3 (NAD+), gamma; isocitrate dehydrogenase 3 gamma; isocitrate dehydrogenase 3, gamma; isocitrate dehydrogenase, NAD(+)-specific, mitochondrial, gamma subunit; isocitric dehydrogenase subunit gamma; NAD (H)-specific isocitrate dehydrogenase gamma subunit; NAD(+)-specific ICDH subunit gamma; NAD+-isocitrate dehydrogenase gamma subunit; NAD+-specific ICDH
Common Name IDH3G
Gene Symbol Idh3g
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence FDVMVMPNLYGNIVNNVCAGLVGGPGLVAGANYGHVYAVFETATRNTGKSIANKNIANPTATLLASCMMLDHLKLHSYATSIRKAVLASMDNENMHTPDIGGQGTTSEAIQDVI
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.