missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ID2 Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems AF4660
This item is not returnable.
View return policy
Description
ID2 Polyclonal specifically detects ID2 in Human samples. It is validated for Western Blot, Immunocytochemistry.
Specifications
| ID2 | |
| Polyclonal | |
| Unconjugated | |
| Q02363 | |
| ID2 | |
| E. coli-derived recombinant human ID2 Met1-Gly134 Accession # Q02363 | |
| 100 μg | |
| Primary | |
| Detects human ID2 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) ID1, rhID3, and rhID4 is observed. | |
| Human | |
| IgG |
| Western Blot, Immunocytochemistry | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| BHLHB26, bHLHb26cell growth-inhibiting gene 8, Class B basic helix-loop-helix protein 26, DNA-binding protein inhibitor ID2, DNA-binding protein inhibitor ID-2, GIG8, helix-loop-helix protein ID2, ID2A, ID2H, inhibitor of differentiation 2, Inhibitor of DNA binding 2, inhibitor of DNA binding 2, dominant negative helix-loop-helix protein, MGC26389 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3398 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction