missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ICOS Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB6975
This item is not returnable.
View return policy
Description
ICOS Monoclonal specifically detects ICOS in Human samples. It is validated for Flow Cytometry, CyTOF-ready.
Specifications
| ICOS | |
| Monoclonal | |
| LYOPH | |
| Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
| Q9Y6W8 | |
| ICOS | |
| NS0 mouse myeloma cell line transfected with human ICOS and mouse myeloma cell line NS0-derived recombinant human ICOS cocktail Glu21-Phe141 Accession # Q9Y6W8 | |
| 100 μg | |
| Primary | |
| Detects human ICOS in direct ELISAs. | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF | |
| 669222 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Activation-inducible lymphocyte immunomediatory molecule, AILIM, AILIMCD278, CD278, CD278 antigen, CRP-1, CVID1, inducible costimulator, inducible T-cell costimulator, inducible T-cell co-stimulator, MGC39850 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 29851 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction