missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human ICMT (aa 239-278) Control Fragment Recombinant Protein

Product Code. 30198276
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30198276

Brand: Invitrogen™ RP95615

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (98%), Rat (98%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-56904 (PA5-56904. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene encodes the third of three enzymes that post translationally modify isoprenylated C-terminal cysteine residues in certain proteins and target those proteins to the cell membrane. This enzyme localizes to the endoplasmic reticulum. Alternative splicing may result in other transcript variants, but the biological validity of those transcripts has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O60725
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 23463
Name Human ICMT (aa 239-278) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 1700008E11Rik; C80758; farnesyl cysteine carboxyl methyltransferase; fcmt; Gm13095; HSTE14; Icmt; isoprenylcysteine carboxyl methyltransferase; Isoprenylcysteine carboxylmethyltransferase; MGC39955; MST098; MSTP098; OTTMUSG00000010406; pcCMT; PCMT; PPMT; prenylated protein carboxyl methyltransferase; prenylcysteine carboxyl methlytransferase; prenylcysteine carboxyl methyltransferase; protein-S isoprenylcysteine O-methyltransferase; protein-S-isoprenylcysteine O-methyltransferase; STE14
Common Name ICMT
Gene Symbol Icmt
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence TVWRFFRDRTEEEEISLIHFFGEEYLEYKKRVPTGLPFIK
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.