missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human ICEBERG (aa 48-90) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (47%), Rat (47%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-58451 (PA5-58451. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Inhibits generation of IL-1-beta by interacting with caspase-1 and preventing its association with RIP2. Down-regulates the release of IL1B.
Specifications
Specifications
| Accession Number | P57730 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 59082 |
| Name | Human ICEBERG (aa 48-90) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | CARD18; caspase recruitment domain family member 18; caspase recruitment domain family, member 18; caspase recruitment domain-containing protein 18; Caspase-1 inhibitor Iceberg; ICEBERG; ICEBERG caspase-1 inhibitor; pseudo-ICE; UNQ5804; UNQ5804/PRO19611 |
| Common Name | ICEBERG |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction