missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ICAM-5 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB1950
This item is not returnable.
View return policy
Description
ICAM-5 Monoclonal specifically detects ICAM-5 in Human samples. It is validated for ELISA Capture (Matched Antibody Pair).
Specifications
| ICAM-5 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| Q9UMF0 | |
| ICAM5 | |
| Mouse myeloma cell line NS0-derived recombinant human ICAM-5 Ala28-Glu570 Accession # Q9UMF0 | |
| 100 μg | |
| Primary | |
| Detects human ICAM-5 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human (rh) CD31, rhCEACAM-1, recombinant mouse (rm) DCC, rhICAM-1, -2, -3, rmICAM-5, rhMAdCAM-1, or rmVCAM-1 is observed. | |
| Human | |
| Purified |
| Western Blot, Immunocytochemistry | |
| 283607 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ICAM5, ICAM-5, intercellular adhesion molecule 5, intercellular adhesion molecule 5, telencephalin, Telencephalin, TLCNTelencephalin, TLNtelencephalin | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 7087 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction