missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ICAM-3/CD50 Antibody, R&D Systems™
Mouse Monoclonal Antibody
£156.00 - £395.00
Specifications
| Antigen | ICAM-3/CD50 |
|---|---|
| Clone | 76203 |
| Dilution | Flow Cytometry 0.25 ug/10^6 cells, Immunohistochemistry 5-25 ug/mL, CyTOF-ready |
| Applications | Flow Cytometry, Immunohistochemistry, CyTOF |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18183371
|
R&D Systems
MAB8131-SP |
25 μg |
£156.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18119320
|
R&D Systems
MAB8131-100 |
100 μg |
£395.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ICAM-3/CD50 Monoclonal antibody specifically detects ICAM-3/CD50 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOFSpecifications
| ICAM-3/CD50 | |
| Flow Cytometry 0.25 ug/10^6 cells, Immunohistochemistry 5-25 ug/mL, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| CD50, CD50 antigen, CDW50CDw50, ICAM3, ICAM-3, ICAM-Rintercellular adhesion molecule-3, intercellular adhesion molecule 3 | |
| Mouse myeloma cell line NS0-derived recombinant human ICAM-3/CD50, Gln30-His485, Accession # CAA49473 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 76203 | |
| Flow Cytometry, Immunohistochemistry, CyTOF | |
| Unconjugated | |
| Mouse | |
| Human | |
| CAA49473 | |
| 3385 | |
| IgG1 | |
| Protein A or G purified from ascites | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 6 months, -20°C to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title