missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ICAM-3/CD50 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF813
This item is not returnable.
View return policy
Description
ICAM-3/CD50 Polyclonal specifically detects ICAM-3/CD50 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.
Specifications
| ICAM-3/CD50 | |
| Polyclonal | |
| Unconjugated | |
| CAA49473 | |
| ICAM3 | |
| Mouse myeloma cell line NS0-derived recombinant human ICAM-3/CD50 Gln30-His485 Accession # CAA49473 | |
| 100 μg | |
| Primary | |
| Detects human ICAM-3/CD50 in direct ELISAs and Western blots. In Western blots, approximately 20% cross-reactivity with recombinant human ICAM-1 is observed. | |
| Human | |
| IgG |
| Western Blot, Flow Cytometry, CyTOF | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
| CD50, CD50 antigen, CDW50CDw50, ICAM3, ICAM-3, ICAM-Rintercellular adhesion molecule-3, intercellular adhesion molecule 3 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3385 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction