missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human ICAM-1 (CD54) Recombinant Protein
Description
This product contains purified recombinant human ICAM-1 protein with an N-terminal hexahistidine tag. This product may be used as a positive control in many immunological assays.
ICAM-1 (CD54) is an 85-110 kDa single-chain type 1 integral membrane glycoprotein with an extracellular domain of five immunoglobulin superfamily repeats, a transmembrane region and a cytoplasmic domain. ICAM-1 has 7 potential N-linked glycosylation sites and shares considerable amino acid sequence homology with ICAM-3 (CD50) and ICAM-2 (CD102). ICAM-1 binds to integrins of type CD11a/CD18 (leukocyte adhesion molecule, LFA-1), or CD11b/CD18 (Mac-1) and is exploited by Rhinovirus as a receptor. ICAM-1 is expressed by activated endothelial cells and detected on epithelial cells, fibroblasts, chondrocytes, B lymphocytes, T lymphocytes (low), monocytes, macrophages, dendritic cells and neutrophils, with lower levels that increase upon inflammation. ICAM-1 is also detected in some carcinoma and melanoma cells. Soluble ICAM-1 is detectable in the plasma and is elevated in patients with various inflammatory syndromes.
Specifications
Specifications
| Accession Number | P05362 |
| Concentration | 1 mg/mL |
| For Use With (Application) | Control, Western Blot |
| Formulation | 0.02 M tris HCl with 50% glycerol and no preservative; pH 8 |
| Gene ID (Entrez) | 3383 |
| Name | Human ICAM-1 (CD54) |
| Quantity | 100 μg |
| Source | E. coli |
| Immunogen | N/A |
| Regulatory Status | RUO |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?