missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ICAM-1/CD54 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 3 publications
Brand: R&D Systems MAB720-SP
This item is not returnable.
View return policy
Description
ICAM-1/CD54 Monoclonal specifically detects ICAM-1/CD54 in Human samples. It is validated for Western Blot, Immunocytochemistry, ELISA Capture (Matched Antibody Pair).
Specifications
| ICAM-1/CD54 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ICAM1 | |
| Chinese hamster ovary cell line CHO-derived recombinant human ICAM-1 Extracellular domain | |
| 25 μg | |
| Primary | |
| Detects recombinant human (rh)ICAM-1 in direct ELISAs and Western blots. In Western blots, shows no cross-reactivity with rrICAM-1, rhICAM-2, rhICAM-3, rhVCAM-1, and rmDCC. | |
| Human | |
| Purified |
| Western Blot, Immunocytochemistry, ELISA | |
| 14C11 | |
| Western Blot 1 ug/mL, Immunocytochemistry 5-25 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| BB2, CD54, CD54 antigen, cell surface glycoprotein P3.58, human rhinovirus receptor, ICAM1, ICAM-1, intercellular adhesion molecule 1, intercellular adhesion molecule 1 (CD54), human rhinovirus receptor, Major group rhinovirus receptor, P3.58 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3383 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction