missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IBSP/Sialoprotein II Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems AF4014-SP
This item is not returnable.
View return policy
Description
IBSP/Sialoprotein II Polyclonal specifically detects IBSP/Sialoprotein II in Human samples. It is validated for Western Blot.
Specifications
| IBSP/Sialoprotein II | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| BNSP, Bone sialoprotein, Bone sialoprotein 2, BSP, BSP 2, BSP II, BSP2, BSPII, BSP-II, Cell binding sialoprotein, IBSP, Integrin binding sialoprotein, SP II, SPII, SP-II | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3381 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| AAC95490 | |
| IBSP | |
| Chinese hamster ovary cell line CHO-derived recombinant human IBSP/Sialoprotein II Phe17-Gln317 Accession # AAC95490 | |
| 25 μg | |
| Primary | |
| Detects human IBSP/Sialoprotein II in direct ELISAs and Western blots. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction