missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human I-309 (CCL1) Recombinant Protein, PeproTech®

Product Code. 17802333
Click to view available options
Quantity:
1 mg
10 μg
100 μg
2 μg
250 μg
500 μg
Unit Size:
100µg
10µg
1mg
250µg
2µg
500µg
This item is not returnable. View return policy

Product Code. 17802333

Brand: Gibco™ 3003710UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Recombinant Human I-309 is an 8.5 kDa protein consisting of 74 amino acid residues that include six cysteine residues, of which four are the highly conserved cysteine residues present in CC chemokines. Unlike some CC chemokines, whose biologically active form in solution is a non-disulfide-linked dimer, the biologically active form of I-309 is monomeric. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

This gene is one of several cytokine genes clustered on the q-arm of chromosome 17. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The protein encoded by this gene is structurally related to the CXC subfamily of cytokines. Members of this subfamily are characterized by two cysteines separated by a single amino acid. This cytokine is secreted by activated T cells and displays chemotactic activity for monocytes but not for neutrophils. It binds to the chemokine receptor CCR8.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P22362
For Use With (Application) ELISA, Functional Assay, Western Blot
Formulation protein with no preservative
Gene ID (Entrez) 6346
Molecular Weight (g/mol) 8.5 kDa
Name Human I-309 (CCL1)
Quantity 10 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias BF534335; C-C motif chemokine 1; C-C motif chemokine ligand 1; CCL 1; Ccl1; CCR8 ligand; chemokine (C-C motif) ligand 1; I-309; inflammatory cytokine I-309; P500; SCYA1; SISe; SIS-epsilon; small inducible cytokine A1; small inducible cytokine A1 (I-309, homologous to mouse Tca-3); small-inducible cytokine A1; T lymphocyte-secreted protein I-309; TCA3; TCA-3; T-cell activation protein 3
Common Name CCL1
Gene Symbol CCL1
Biological Activity Determined by its ability to chemoattract human monocytes population using a concentration range of 10.0-30.0 ng/ml
Conjugate Unconjugated
Recombinant Recombinant
Sequence SKSMQVPFSR CCFSFAEQEI PLRAILCYRN TSSICSNEGL IFKLKRGKEA CALDTVGWVQ RHRKMLRHCP SKRK
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.