Learn More
Gibco™ Human HVEM Fc Recombinant Protein, PeproTech®
Description
310-27-1MG will be provided as 10 x 100 μg (310-27-100UG). 310-27-200UG will be provided as 2 x 100 μg (310-27-100UG). 310-27-500UG will be provided as 5 x 100 μg (310-27-100UG). Recombinant Human HVEM-Fc Chimera is a 376 amino acid fusion protein that contains an N-terminal domain corresponding to the extracellular region of HVEM, and a C-terminal domain corresponding to residues 102 to 330 of human IgG1. The calculated molecular weight of Recombinant Human HVEM-Fc is 41.4 kDa. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
Specifications
Specifications
| Accession Number | Q92956 |
| For Use With (Application) | Functional Assay |
| Formulation | protein with no preservative |
| Gene ID (Entrez) | 8764 |
| Molecular Weight (g/mol) | 41.4 |
| Name | Human HVEM Fc |
| Quantity | 5 x 100 μg |
| Source | Insect cells |
| Storage Requirements | -20°C |
| Regulatory Status | RUO |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.