missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human HTRA3 (aa 337-449) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Conditionnement:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (89%), Rat (89%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-110869 (PA5-110869. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
HtrA3 belongs to the human high-temperature requirement factor A (HtrA) family. It is also called pregnancy-related serine protease. HtrA3 has two isoforms (long and short), which is produced by alternative splicing. Similar to HtrA1, HtrA3 binds to various TGF-beta proteins and inhibited the signaling of BMP-4, -2 and TGF-beta 1. HtrA3 does not inhibit signaling originated from a constitutively active BMP receptor, indicating that the inhibition occurred upstream of the cell surface receptor.
Spécification
Spécification
| Accession Number | P83110 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 94031 |
| Name | Human HTRA3 (aa 337-449) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | 2210021K23Rik; 9530081K03Rik; High-temperature requirement factor A3; HtrA serine peptidase 3; Htra3; Pregnancy-related serine protease; probable serine protease HTRA3; Prsp; RGD1308120; Serine protease HTRA3; Tasp; Toll-associated serine protease |
| Common Name | HTRA3 |
| Afficher plus |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu