Learn More
Abcam Human HTR3D (5HT3D Receptor) knockout A549 cell line
Product Code. 32139319 Shop All Abcam Products
Change view
Click to view available options
Quantity:
2 x 1,000,000 Cells
Unit Size:
Pack of 2
Product Code. Quantity Unit Size
32139319 2 x 1,000,000 Cells Pack of 2
1 options
Product Code. 32139319 Supplier Abcam Supplier No. ab2669922x1000000Cells_
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
HTR3D KO cell line available to order. KO validated. Free of charge wild type control available. Knockout achieved by using CRISPR/Cas9, 1 bp deletion in exon 6 and 2 bp insertion in exon 6. To order both knockout and wild-type control cells: select 2 x 1000000 Cells/vial. To order only knockou...
HTR3D KO cell line available to order. KO validated. Free of charge wild type control available. Knockout achieved by using CRISPR/Cas9, 1 bp deletion in exon 6 and 2 bp insertion in exon 6. To order both knockout and wild-type control cells: select 2 x 1000000 Cells/vial'. To order only knockout cells: select 1000000 Cells/vial'.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Quantity 2 x 1,000,000 Cells