missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human HSPB9 (aa 80-159) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (55%), Rat (55%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-144846 (PA5-144846. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
HSPB9 gene ontology (GO) annotation includes protein binding and nucleus.
Specifications
Specifications
| Accession Number | Q9BQS6 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 94086 |
| Name | Human HSPB9 (aa 80-159) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | 1700007H20Rik; Cancer/testis antigen 51; CT51; Heat shock protein; Heat shock protein beta-9; heat shock protein family B (small) member 9; heat shock protein, alpha-crystallin-related, B9; HSP; HSPB9; small heat shock protein B9 |
| Common Name | HSPB9 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction