missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human HSPA4L (aa 539-600) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (85%), Rat (85%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-58671 (PA5-58671. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
HSPA4L possesses chaperone activity in vitro where it inhibits aggregation of citrate synthase.
Specifications
Specifications
| Accession Number | O95757 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 22824 |
| Name | Human HSPA4L (aa 539-600) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | 94 kDa; AI461691; APG1; APG-1; heat shock 70 kDa protein 4 L; heat shock 70 kDa protein 4-like protein; heat shock 70 kDa protein 4 L-like; Heat shock 70 kDa protein 4 L-like protein; heat shock 70 kDa protein 4 like; heat shock 70 kDa protein 4-like; heat shock 70-related protein APG-1; Heat shock protein; heat shock protein (hsp110 family); heat shock protein 4 like; heat shock protein 4-like; heat shock protein family A (Hsp70) member 4 like; HSP; Hsp4l; HSPA4L; HSPH3; LOW QUALITY PROTEIN: heat shock 70 kDa protein 4 L; osmotic stress protein 94; OSP94; testis tissue sperm-binding protein Li 64 n |
| Common Name | HSPA4L |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction