missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human HPS1 (aa 374-458) Control Fragment Recombinant Protein

Product Code. 30210251
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30210251

Brand: Invitrogen™ RP101171

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (75%), Rat (75%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-63938 (PA5-63938. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Hermansky-Pudlak syndrome (HPS) is a rare, genetically heterogeneous, autosomal recessive disorder. It is characterized by oculocutaneous albinism, lysosomal storage defects and prolonged bleeding due to platelet storage pool deficiency. There are 10 HPS genes encoding HPS proteins that all interact within three distinct ubiquitously expressed protein complexes or biogenesis of lysosome-related organelle complexes. Defects in these genes cause HPS. HSP-1 is a component of multiple cytoplasmic organelles and is important for their normal development and function. It plays a role in intracellular protein sorting.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q92902
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 3257
Name Human HPS1 (aa 374-458) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 6030422N11Rik; BB405864; BLOC3S1; ep; Gm21361; Hermansky-Pudlak syndrome 1 homolog (human); Hermansky-Pudlak syndrome 1 protein; Hermansky-Pudlak syndrome 1 protein homolog; Hermansky-Pudlak syndrome protein homolog; HPS; HPS1; HPS1, biogenesis of lysosomal organelles complex 3 subunit 1; MGC5277; pale ear
Common Name HPS1
Gene Symbol HPS1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence LWQGINLVLLTRSPSAPLALVLSQLMDGFSMLEKKLKEGPEPGASLRSQPLVGDLRQRMDKFVKNRGAQEIQSTWLEFKAKAFSK
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.