missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human HPRG Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB1869-SP
Denne vare kan ikke returneres.
Se returpolicy
Beskrivelse
HPRG Monoclonal specifically detects HPRG in Human samples. It is validated for Western Blot.
Tekniske data
| HPRG | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| P04196 | |
| HRG | |
| Mouse myeloma cell line NS0-derived recombinant human HPRG Val19-Lys525 Accession # P04196 | |
| 25 μg | |
| Primary | |
| Detects human HPRG in direct ELISAs and Western blots. In direct ELISAs, this antibody does not cross-react with recombinant human (rh) Cystatin B, C, D, E/M, S, A, SA, SN, rhFetuin A, B, rmHPRG, rhKininogen, or rhKininostatin. | |
| Human | |
| Purified |
| Western Blot | |
| 227901 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| DKFZp779H1622, Histidine-proline-rich glycoprotein, histidine-rich glycoprotein, HPRGHRGP, HRG | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3273 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
For Research Use Only
Ser du en mulighed for forbedring?Del en indholdskorrektion