missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human HPRG Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB1869
This item is not returnable.
View return policy
Description
HPRG Monoclonal specifically detects HPRG in Human samples. It is validated for Western Blot.
Specifications
| HPRG | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| P04196 | |
| HRG | |
| Mouse myeloma cell line NS0-derived recombinant human HPRG Val19-Lys525 Accession # P04196 | |
| 500 μg | |
| Primary | |
| Detects human HPRG in direct ELISAs and Western blots. In direct ELISAs, this antibody does not cross-react with recombinant human (rh) Cystatin B, C, D, E/M, S, A, SA, SN, rhFetuin A, B, rmHPRG, rhKininogen, or rhKininostatin. | |
| Human | |
| Purified |
| Western Blot | |
| 227901 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| DKFZp779H1622, Histidine-proline-rich glycoprotein, histidine-rich glycoprotein, HPRGHRGP, HRG | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3273 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction