missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human HPa2 Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (96%), Rat (96%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-60773 (PA5-60773. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
This gene encodes a heparanase enzyme. The encoded protein is a endoglycosidase that degrades heparin sulfate proteoglycans located on the extracellular matrix and cell surface. This protein may be involved in biological processes involving remodeling of the extracellular matrix including angiogenesis and tumor progression. Alternate splicing results in multiple transcript variants.
Specifications
Specifications
| Accession Number | Q8WWQ2 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 60495 |
| Name | Human HPa2 Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | Gm968; heparanase 2; heparanase 2 (inactive); heparanase 3; heparanase-2; heparanase-like protein; Hpa2; HPR2; Hpse2; Inactive heparanase-2; UFS; UFS1 |
| Common Name | HPa2 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction