Learn More
Abnova™ Human HOXC8 Full-length ORF (NP_073149.1, 1 a.a. - 242 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00003224-P01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene belongs to the homeobox family of genes. The homeobox genes encode a highly conserved family of transcription factors that play an important role in morphogenesis in all multicellular organisms. Mammals possess four similar homeobox gene clusters, HOXA, HOXB, HOXC and HOXD, which are located on different chromosomes and consist of 9 to 11 genes arranged in tandem. This gene is one of several homeobox HOXC genes located in a cluster on chromosome 12. The product of this gene may play a role in the regulation of cartilage differentiation. It could also be involved in chondrodysplasias or other cartilage disorders. [provided by RefSeq]
Sequence: MSSYFVNPLFSKYKAGESLEPAYYDCRFPQSVGRSHALVYGPGGSAPGFQHASHHVQDFFHHGTSGISNSGYQQNPCSLSCHGDASKFYGYEALPRQSLYGAQQEASVVQYPDCKSSANTNSSEGQGHLNQNSSPSLMFPWMRPHAPGRRSGRQTYSRYQTLELEKEFLFNPYLTRKRRIEVSHALGLTERQVKIWFQNRRMKWKKENNKDKLPGARDEEKVEEEGNEEEEKEEEEKEENKDSpecifications
NP_073149.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
54.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HOX3/HOX3A | |
HOXC8 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
3224 | |
HOXC8 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MSSYFVNPLFSKYKAGESLEPAYYDCRFPQSVGRSHALVYGPGGSAPGFQHASHHVQDFFHHGTSGISNSGYQQNPCSLSCHGDASKFYGYEALPRQSLYGAQQEASVVQYPDCKSSANTNSSEGQGHLNQNSSPSLMFPWMRPHAPGRRSGRQTYSRYQTLELEKEFLFNPYLTRKRRIEVSHALGLTERQVKIWFQNRRMKWKKENNKDKLPGARDEEKVEEEGNEEEEKEEEEKEENKD | |
RUO | |
HOXC8 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |