missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human HOXB9 Partial ORF (NP_076922.1, 65 a.a. - 163 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00003219-Q01.10ug
This item is not returnable.
View return policy
Description
This gene is a member of the Abd-B homeobox family and encodes a protein with a homeobox DNA-binding domain. It is included in a cluster of homeobox B genes located on chromosome 17. The encoded nuclear protein functions as a sequence-specific transcription factor that is involved in cell proliferation and differentiation. Increased expression of this gene is associated with some cases of leukemia, prostate cancer and lung cancer. [provided by RefSeq]
Sequence: PLSPHASGSLPSVYHPYIQPQGVPPAESRYLRTWLEPAPRGEAAPGQGQAAVKAEPLLGAPGELLKQGTPEYSLETSAGREAVLSNQRPGYGDNKICEGSpecifications
NP_076922.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.52kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
PLSPHASGSLPSVYHPYIQPQGVPPAESRYLRTWLEPAPRGEAAPGQGQAAVKAEPLLGAPGELLKQGTPEYSLETSAGREAVLSNQRPGYGDNKICEG | |
RUO | |
HOXB9 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
3219 | |
HOXB9 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HOX-2.5/HOX2/HOX2E | |
HOXB9 | |
Recombinant | |
wheat germ expression system |