missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human HOXB7 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB80401
This item is not returnable.
View return policy
Description
HOXB7 Monoclonal specifically detects HOXB7 in Human samples. It is validated for Western Blot.
Specifications
| HOXB7 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2 ug/mL | |
| P09629 | |
| HOXB7 | |
| E. coli-derived recombinant human HOXB7 Ser2-Asn123 Accession # P09629 | |
| 100 μg | |
| Primary | |
| Detects human HOXB7 in direct ELISAs and Western blots. In direct ELISAa, no cross-reactivity with recombinant human HOXA7 is observed. | |
| Human | |
| Purified |
| Western Blot | |
| 864617 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| HHO.C1, homeo box 2C, homeo box B7, homeo box c1 protein, homeobox B7, Homeobox protein HHO.C1, Homeobox protein Hox-2C, homeobox protein Hox-B7, HOX2, Hox-2.3, HOX2C, HOX2CHox-2.3 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3217 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction